Monoammonium glycyrrhizinate hydrate ≥98%

Supplier: Spectrum Chemical Mfg. Corp.

G1403-25GM G1403-100GM
700009-504EA 177.75 USD
700009-504 700009-502
Monoammonium glycyrrhizinate hydrate ≥98%
Monoammonium glycyrrhizinate hydrate

Glycyrrhizic Acid Monoammonium Salt, Trihydrate is the primary cause of the sweet flavor of the licorice root. It has also been used as a treatment for hepatitis C in Japan and as an emulsifier in cosmetics and certain foods. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Formula: C₄₂H₆₅NO₁₆·xH₂O
Storage Temperature: Ambient
MDL Number: MFCD00167400
CAS Number: 53956-04-0

Order Now

Specification Test Results

Assay (C₄₂H₆₅NO₁₆.3H₂O) 98 % min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR