4-Hydroxy-3-methoxycinnamic acid (Ferulic acid), powder
Supplier: SPECTRUM CHEMICAL MFG CORP
|
|
Synonyms:
3-(4-Hydroxy-3-methoxyphenyl)acrylic acid, NSC 2821, NSC 674320, NSC 51986, Coniferic acid, Ferulic acid
F1106-100GM
F1106-25GM
F1106-1KGBL
F1106-5GM
700014-294EA
605.75
USD
700014-294
700014-298
700014-296
700014-300
4-Hydroxy-3-methoxycinnamic acid (Ferulic acid), powder
4-Hydroxy-3-methoxycinnamic acid (Ferulic acid)
Ferulic Acid, Powder is also known as a 3-methoxy-4-hydroxycinnamic acid and is an organic compound that is found naturally occuring in nature in seeds and cereals. It is a potent antioxidant, a protein matrix in mass spectrometry analysis and as a manufacturing precursor of vanillin.
GHS hazard : Exclamation mark
Formula:
C₁₀H₁₀O₄ Storage Temperature: Ambient |
MDL Number:
MFCD00004400 CAS Number: 1135-24-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...