Iron(II) fumarate ≥97.0% USP

Supplier: Spectrum Chemical Mfg. Corp.

FE108-50KGBL FE108-2.5KG FE108-500GM FE108-12KG
700005-924EA 1560.85 USD
700005-924 95033-230 95033-232 700005-922
Iron(II) fumarate ≥97.0% USP
Iron(II) fumarate

Ferrous Fumarate, USP is a mineral supplement. 


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₄H₂FeO₄
Storage Temperature: Ambient
MDL Number: MFCD00058315
CAS Number: 141-01-5

Order Now

Specification Test Results

Assay (C₄H₂Feo₄) 97.0-101.0 %
Loss On Drying max.1.5 %
Sulfates (SO₄) max. 0.2 %
Arsenic (As) max. 3 ppm
Ferric Iron (Fe [D) max. 2.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR