Etidronic acid 60%
Supplier: Spectrum Chemical Mfg. Corp.
|
|
Etidronic Acid, also known as 1-hydroxyethane 1,1-diphosphonic acid, is used as a bisphosphonate to treat osteoporosis. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
GHS hazard : Exclamation mark;Corrosion
Formula:
C₂H₈O₇P₂ Storage Temperature: Ambient |
MDL Number:
MFCD00070585 CAS Number: 2809-21-4 UN: 3261 ADR: 8,III |
Specification Test Results
Assay (In Water) | Approx. 60 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...