Erioglaucine disodium salt

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Sodium 2-((4-(ethyl(3-sulfonatobenzyl)amino)phenyl)(4-(ethyl(3-sulfonatobenzyl)iminio)cyclohexa-2,5-dien-1-ylidene)methyl)benzenesulfonate, FD and C Blue No. 1, Brilliant Blue FCF, Erioglaucin A, Sodium 2-((4-(ethyl(3-sulphonatobenzyl)amino)phenyl)(4-(ethyl(3-sulphonatobenzyl)iminio)cyclohexa-2,5-dien-1-ylidene)methyl)benzenesulphonate, Alphazurine FG, Erioglaucine sodium salt, CI 42090, Acid blue 9, Food Blue No.1

ER120-5GM ER120-25GM
700010-038EA 57.7 USD
700010-038 700010-036
Erioglaucine disodium salt
Erioglaucine disodium salt

Erioglaucine, also known as Brilliant Blue FCF, is commonly a colorant for foods like ice cream, processed peas, and dairy products. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Formula: C₃₇H₃₄Na₂N₂O₉S₃
MW: 792.84 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012141
CAS Number: 3844-45-9

Order Now

Specification Test Results

Absorption Max 627.0 - 631.0 NM


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR