D-Saccharic acid calcium salt
Supplier: Spectrum Chemical Mfg. Corp.
|
|
D-Saccharic Acid Calcium Salt is used to study its potential chemopreventive properties and anti-oxidative properties at the level of peroxynitrite-induced damages.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
CaC₆H₈O₈·xH₂O Storage Temperature: Ambient |
MDL Number:
MFCD00043409 CAS Number: 5793-88-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...