D-Mannose 6-phosphate sodium salt ≥95%

Supplier: Spectrum Chemical Mfg. Corp.

M3065-100MG M3065-1GM
700009-672EA 262.65 USD
700009-672 700009-674
D-Mannose 6-phosphate sodium salt ≥95%
D-Mannose 6-phosphate sodium salt

Storage temperature - REFRIGERATE: Keep tightly closed.

Storage Temperature: Refrigerator
MDL Number: MFCD00135859
CAS Number: 70442-25-0

Order Now

Specification Test Results

Assay Approx. 95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR