Diethylenetriamine ≥98%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 2,2'-Iminodi(ethylamine), DIEN, Diethylenetriamine

D1092-1LTGL D1092-20LT D1092-4LTGL D1092-500MLGL
10841-778EA 143.7 USD
10841-778 10841-780 10841-782 10841-784
Diethylenetriamine ≥98%
Diethylenetriamine

Diethylenetriamine is also known under the abbreviation DETA and is a colorless liquid that is hygroscopic and soluble in both water and polar organic solvents. Used as a ligand forming complex in coordination chemistry, it is also used for the extraction of gases that are acid in the oil industry.


GHS hazard : Corrosion;Exclamation mark;Health hazard UN2079

Formula: C₄H₁₃N₃
MW: 103.17 g/mol
Boiling Pt: 207
Melting Pt: -39
Density: 0.955
Flash Pt: 97
Storage Temperature: Ambient
MDL Number: MFCD00008171
CAS Number: 111-40-0
UN: 2079
ADR: 8 II

Order Now

Specification Test Results

Assay 98 % min.
Water max. 0.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR