Diethyl oxalate ≥99.0%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Oxalic acid diethyl ester

D2365-2.5LTPV D2365-500MLGL D2365-100ML
10841-772EA 78.54 USD
10841-772 10841-776 10841-774
Diethyl oxalate ≥99.0%
Diethyl oxalate

Caution: Storage temperature - HYGROSCOPIC: Keep tightly closed.

Formula: C₆H₁₀O₄
MW: 146.14 g/mol
Density: 1.076
Storage Temperature: Ambient
MDL Number: MFCD00009119
CAS Number: 95-92-1
UN: 2525
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99.0 % min.
Color (APHA) max.20
Residue On Ignition max. 0.005 %
Water max. 0.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR