Quinoline yellow sulfonic acid sodium salt (Acid yellow 3)

Supplier: Spectrum Chemical Mfg. Corp.

DC200-100GM DC200-25GM DC200-5KGBL DC200-500GM
700008-756EA 483 USD
700008-756 700008-758 700008-762 700008-760
Quinoline yellow sulfonic acid sodium salt (Acid yellow 3)
Quinoline yellow sulfonic acid sodium salt (Acid yellow 3)

D and C Yellow No. 10 | Non-graded D and C Yellow No. 10 supplied by Spectrum is indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.


GHS hazard : Exclamation mark

Formula: C₁₈H₁₁NO₂.xNaO₃S
Storage Temperature: Ambient
MDL Number: MFCD00080727
CAS Number: 8004-92-0

Order Now

Specification Test Results

Arsenic (As) To Pass Test
Lead (Pb) To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR