Pigment red 57 ≥90%

Supplier: Spectrum Chemical Mfg. Corp.

D1139-500GM D1139-100GM D1139-25GM
700013-996EA 1472.1 USD
700013-996 700013-992 700013-994
Pigment red 57 ≥90%
Pigment red 57

D and C Red No. 6 | Non-graded D and C Red No. 6 supplied by Spectrum is indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₈H₁₄N₂O₆S·₂Na
Storage Temperature: Ambient
MDL Number: MFCD04974896
CAS Number: 5858-81-1

Order Now

Specification Test Results

Assay 90% min.
Identification To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR