Copper(II) sulfate pentahydrate ≥98.5%, powder USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Cupric sulfate pentahydrate, Cupric sulphate pentahydrate, Copper sulphate pentahydrate, Copper sulfate pentahydrate

CU110-2.5KG CU110-125GM CU110-12KG CU110-500GM CU110-45KGBL
95030-676EA 851 USD
95030-676 95030-674 89078-780 95030-678 75810-954
Copper(II) sulfate pentahydrate ≥98.5%, powder USP
Copper (II) sulphate pentahydrate

Cupric Sulfate, Pentahydrate, Powder, USP is a mineral supplement. 


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: CuSO₄·5H₂O
MW: 249.69 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149681
CAS Number: 7758-99-8
UN: 3077
ADR: 9 III
Merck Index: 13,02682

Order Now

Specification Test Results

Assay (Dried Basis) 98.5 - 100.5%
Loss On Drying 33.0 - 36.5%
Sodium (Na) max. 0.02%
Potassium (K) max. 0.01%
Calcium (Ca) max. 0.005%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR