Cromolyn sodium ≥98.0% USP
|
|
Synonyms:
Disodium Cromoglycate
, Sodium 5,5'-((2-hydroxypropane-1,3-diyl)bis(oxy))bis(4-oxo-4H-chromene-2-carboxylate)
, 5,5'-[(2-Hydroxy-1,3-propanediyl)bis(oxy)]bis(4-oxo-4H-1-benzopyran-2-carboxylic Acid) Disodium Salt
C1360-25GM
C1360-100GM
C1360-25KGBL
C1360-5GM
C1360-1GM
C1360-500GMB
95032-726EA
541.62
USD
95032-726
95032-718
700006-274
95032-720
95032-724
95032-722
Cromolyn sodium ≥98.0% USP
Cromolyn sodium
Cromolyn Sodium, USP is an anti-inflammatory medication. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities
GHS hazard : Exclamation mark
Formula:
C₂₃H₁₆O₁₁ MW: 512.33 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00057744 CAS Number: 15826-37-6 |
Specification Test Results
Assay (Anhydrous Basis) | 98.0 - 101.0 % |
Acidity or Alkalinity | To Pass Test |
Water Determination | max. 10.0 % |
Heavy Metals | max. 0.002 % |
Oxalate | max. 0.35 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...