Copal gum, lumps

Supplier: Spectrum Chemical Mfg. Corp.

CO134-500GM CO134-2.5KG
700013-976EA 116 USD
700013-976 700013-974
Copal gum, lumps
Copal gum

Copal Gum, Lump, also known as gum copal, is a resin from the forest tree and can be used in varnishes as a component for carriages.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Storage Temperature: Ambient
CAS Number: 9000-14-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR