Cetyl esters wax (synthetic) NF

Supplier: Spectrum Chemical Mfg. Corp.

SP310-2.5KG SP310-500GM SP310-12KG SP310-45KGBL
95032-482EA 392.4 USD
95032-482 95032-484 89078-710 89078-712
Cetyl esters wax (synthetic) NF
Cetyl esters wax

Cetyl Esters Wax, Synthetic, NF is also known as cetyl palmitate and is the ester from cetyl alcohol and palmitic acid. 


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Storage Temperature: Ambient
CAS Number: 8002-23-1

Order Now

Specification Test Results

Melting Range 43 - 47 °C
Saponification Value 109 - 120
Iodine Value max. 1
Acid Value max. 5
Paraffin and Free Acids To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR