Catalase
Supplier: Spectrum Chemical Mfg. Corp.
C1328-1GM C1328-5GM
700011-376EA 195.42 USD
700011-376 700011-380
Catalase
Enzymes
Catalase, Lyophilized, a tetramer of four polypeptide chains, is used in the food industry to remove hydrogen peroxide from milk in order to make cheese. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.