Carrageenan NF

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Irish Moss

C1107-500GM C1107-5GM C1107-100GM C1107-500GM C1107-12KGBL C1107-25GM
89078-700EA 2373.54 USD
89078-700 95032-452 CA95032-450 95032-450 95032-448 95032-446
Carrageenan NF
Carrageenan

Carrageenan, NF is used as thickening agent in lotion formulations. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Storage Temperature: Ambient
MDL Number: MFCD00081480
CAS Number: 9000-07-1

Order Now

Specification Test Results

Solubility (In H₂0) To Pass Test
Viscosity of Solution 5 cPs
Loss On Drying max. 12.5 %
Microbial Limits To Pass Test
Acid-Insoluble Matter max. 2.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR