Carnauba wax, yellow No. 1
Supplier: Spectrum Chemical Mfg. Corp.
Carnauba Wax, Yellow No. 1, Flakes, also called palm wax or Brazil wax, is derived from the leaves of the palm plant. Carnauba wax can produce a glossy finish when used in automobile waxes, shoe polishes, dental floss, food products.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Storage Temperature:
Ambient |
MDL Number:
MFCD00130724 CAS Number: 8015-86-9 |
Specification Test Results
Melting Range | 82.0 - 85.5 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...