Carboxymethylcellulose calcium salt
Supplier: SPECTRUM CHEMICAL MFG CORP
C1528-25GM
C1528-500GM
C1528-100GM
700013-772EA
151.3
USD
700013-772
700013-774
700013-770
Carboxymethylcellulose calcium salt
Carboxymethylcellulose calcium salt
Carboxymethylcellulose Calcium Salt, also known as Carboxymethyl cellulose or cellulose gum, can be used as a thickening agent in pharmaceuticals.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Storage Temperature:
Ambient |
MDL Number:
MFCD00197782 CAS Number: 9050-04-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...