Carbomer 934P resin, powder NF

Supplier: Spectrum Chemical Mfg. Corp.

CA184-100GM CA184-2.5KG CA184-500GM
95032-398EA 258.45 USD
95032-398 95032-400 95032-402
Carbomer 934P resin, powder NF
Carbomer 934P

Carbomer 934P, Resin, NF polymers can provide bioadhesion and good binding characteristics that provide highly effective controlled-release properties. 


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Storage Temperature: Ambient
MDL Number: MFCD00084394
CAS Number: 9062-04-8

Order Now

Specification Test Results

Carboxylic Acid Groups (Dried Basis) 56.0 - 68.0 %
Viscosity of a 0.5% Solution 29,400-39,400 mPa·s
Loss On Drying max. 2.0%
Heavy Metals max. 20 µg/g
Benzene max. 0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR