Calcium-D-saccharate tetrahydrate ≥98.5% USP
Supplier: SPECTRUM CHEMICAL MFG CORP
|
|
Synonyms:
Calcium D-glucarate tetrahydrate
C1830-100GM
95032-328EA
486.43
USD
95032-328
Calcium-D-saccharate tetrahydrate ≥98.5% USP
Calcium-D-saccharate tetrahydrate
Calcium Saccharate, Tetrahydrate, USP is used as an antacid, as an antidote in carbolic acid poisoning, and as a stabilizer for calcium gluconate solution for parenteral administration. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
GHS hazard : Exclamation mark
Formula:
C₆H₈CaO₈.4H₂O Storage Temperature: Ambient |
MDL Number:
MFCD00150704 CAS Number: 5793-89-5 |
Specification Test Results
Assay | 98.5 - 102.0% |
Specific Rotation [A]D | +18.5 to +22.5° |
Chloride (Cl) | max. 0.07% |
Sulfate | max. 0.12% |
Heavy Metals | max. 0.002% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...