Brucine sulfate heptahydrate ACS

Supplier: Spectrum Chemical Mfg. Corp.

BR170-100GM BR170-10GM BR170-25GM
700009-612EA 219.57 USD
700009-612 700009-614 700009-618
Brucine sulfate heptahydrate ACS
Brucine sulfate heptahydrate

GHS hazard : Skull and crossbones UN1544

Formula: C₂₃H₂₆N₂O₄
Storage Temperature: Ambient
MDL Number: MFCD00013472
CAS Number: 60583-39-3
UN: 1544
ADR: 6.1,I

Order Now

Specification Test Results

Sensitivity to Nitrate To Pass Test
Clarity of Solution To Pass Test
Loss On Drying @ 105 C max. 13.0 %
Residue After Ignition max. 0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR