6-O-Palmitoyl-L-ascorbic acid ≥95.0% FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: L-Ascorbic acid 6-palmitate, Palmitoyl-L-ascorbic acid, Ascorbyl Palmitate, (S)-2-((R)-3,4-Dihydroxy-5-oxo-2,5-dihydrofuran-2-yl)-2-hydroxyethyl palmitate, L(+)-Ascorbyl palmitate

A1374-125GM A1374-25GM A1374-500GM A1374-5KGBL
95031-876EA 288.89 USD
95031-876 95031-878 95031-880 75810-536
6-O-Palmitoyl-L-ascorbic acid ≥95.0% FCC
6-O-Palmitoyl-L-ascorbic acid

Ascorbyl Palmitate, FCC is used as an antioxidant food additive (E number E304). The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Formula: C₂₂H₃₈O₇
MW: 414.54 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005377
CAS Number: 137-66-6

Order Now

Specification Test Results

Assay (Dried Basis) 95.0 % min.
Melting Range 107 - 117 °C
Specific Rotation (C=10 ; Methanol) +21 to +24
Lead (Pb) max. 2 mg/kg
Loss On Drying max. 2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR