6-O-Palmitoyl-L-ascorbic acid ≥95.0% NF

Supplier: Spectrum Chemical Mfg. Corp.

A1372-25KGBL A1372-5KGBL A1372-25GM A1372-125GM A1372-500GM
700009-356EA 8622.1 USD
700009-356 700009-358 95031-884 95031-882 95031-886
6-O-Palmitoyl-L-ascorbic acid ≥95.0% NF
6-O-Palmitoyl-L-ascorbic acid

Ascorbyl Palmitate, NF is often used to promote flow of tableting powders and to prevent their sticking in tablet presses. 


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Formula: C₂₂H₃₈O₇
Storage Temperature: Ambient
MDL Number: MFCD00005377
CAS Number: 137-66-6

Order Now

Specification Test Results

Assay (Dried Basis) 95.0 - 100.5 %
Specific Rotation [A]D25 +21° to +24°
Loss On Drying max. 2.0 %
Residue On Ignition max. 0.1 %
Heavy Metals max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR