L(+)-Arginine ≥98.5% USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: H-L-Arg-OH, (S)-2-Amino-5-guanidinopentanoic acid, Arginine, (S)-(+)-Arginine, L-Arginine, H-Arg-OH

A1334-1KG A1334-12KGBL A1334-50KGBL A1334-100GM A1334-25GM A1334-25KGBL
95033-852EA 796.35 USD
95033-852 700007-882 700007-884 95033-850 95033-854 75810-522
L(+)-Arginine ≥98.5% USP
L(+)-Arginine

Arginine, USP is a semiessential amino acid. 


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₆H₁₄N₄O₂
MW: 174.2 g/mol
Boiling Pt: 368
Melting Pt: 238
Storage Temperature: Ambient
MDL Number: MFCD00002635
CAS Number: 74-79-3
Merck Index: 13,00785

Order Now

Specification Test Results

Assay (Dried Basis) 98.5 - 101.5 %
Specific Rotation [A]D +26.3 to +27.7
Loss On Drying max. 0.5 %
Residue On Ignition max. 0.3 %
Chloride (Cl) max. 0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR