6-O-α-Maltosyl-β-cyclodextrin

Supplier: Spectrum Chemical Mfg. Corp.

M3801-250MG M3801-1GM
700009-684EA 160.8 USD
700009-684 700009-682
6-O-α-Maltosyl-β-cyclodextrin
6-O-alpha-Maltosyl-beta-cyclodextrin

6-O-alpha-Maltosyl-beta-cyclodextrin is a personal care ingredient used for skin conditioning properties.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₅₄H₉₀O₄₅
Storage Temperature: Ambient
MDL Number: MFCD00671560
CAS Number: 104723-60-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR