1-Methylnaphthalene ≥94.5%
Supplier: Spectrum Chemical Mfg. Corp.
|
|
1-Methylnaphthalene, also known as alpha-methylnaphthalene, is a polycyclic aromatic hydrocarbon with a cetane number of zero.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₁₁H₁₀ Density: 1.001 Storage Temperature: Ambient |
MDL Number:
MFCD00004034 CAS Number: 90-12-0 UN: 3082 ADR: 9,III |
Specification Test Results
Assay | 94.5 % min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...