Sodium thiosulfate pentahydrate

Supplier: AQUA SOLUTIONS MS

8804-20L 8804-4L 8804-1L
101224-430EA 225.12 USD
101224-430 101226-370 101226-602
Sodium thiosulfate pentahydrate
Sodium thiosulphate pentahydrate
Formula: Na₂S₂O₃.5H₂O
MW: 248.19 g/mol
Boiling Pt: 100 °C
Melting Pt: 48
Density: 1.003 g/cm³ (8.37 lbs/gal.)
MDL Number: MFCD00149186
CAS Number: 10102-17-7
Merck Index: 12,08844

Order Now

Specification Test Results

Prepared to formulation on file Yes
Actual Density 1.00258 - 1.00301
Appearance and color Clear water-white liquid
Normality 0.03932 - 0.03948
Pprepared to ASTM D 1510 Yes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR