n-Heptane, high purity

Supplier: AQUA SOLUTIONS MS
Danger

H1903-4L H1903-1L H1903-500ML H1903-20L
101224-678EA 373.64 USD
101224-678 101225-444 101223-476 101225-382
n-Heptane, high purity
n-Heptane
Formula: H₃C(CH₂)₅CH₃
MDL Number: MFCD00009544
CAS Number: 142-82-5
UN: 1206
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ff97126664c92362bf0aa2645719548c99f0533&vid=6722523a326fdf1ebabd08a2b8656dd57300787f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (as n-Heptane) 99.0% Min
Assay (as C7 Hydrocarbons) 99.5% Min.
Assay (Total Hydrocarbons) 99.9% Min
Specific Gravity @ 15.56C 0.680 - 0.695
Titrable Acid 0.0003 meq/g Max
Residue After Evaporation 10 ppm Max.
Sulfur Compounds (as S) 0.005% Max
Water 0.03% Max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR