Petroleum spirit, 35…60 °C

Supplier: AQUA SOLUTIONS MS
Danger

P1926-20L P1926-4L
101204-216EA 412.94 USD
101204-216 101203-238
Petroleum spirit, 35…60 °C
Petroleum spirit
MDL Number: MFCD00081849
CAS Number: 64742-49-0
UN: 1268
ADR: 3,II

Order Now

Specification Test Results

Residue after Evaporation 10 ppm max.
Sulfur 5 ppm max.
Benzene 2 ppm max.
APHA Color 10 max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR