Super-Size AkroBins, Akro-Mils
Supplier: Akro-Mils
AkroBins®
30281BLUE 30281RED 30292MOBYELLO 30293YELLO 30288YELLO 30283GREY 30287RED 30293MOBGREY 30286BLUE 30280GREY 30292MOBRED 30284SCLAR 30282RED 30292BLUE 30289RED 30281GREY 30288MOBBLUE 30286SCLAR 30280YELLO 30281L 30292SCLAR 30286L 30293MOBYELLO 30289SCLAR 30293GREY 30293MOBRED 30287BLUE 30292MOBGREY 30289BLUE 30287SCLAR 30284YELLO 30280BLUE 30289MOBBLUE 30286GREY 30289MOBSCLAR 30283YELLO 30282SCLAR 30288GREY 30281SCLAR 30282GREY 30288RED 30283L 30286RED 30284GREY 30289GREY 30290MOBSCLAR 30290MOBRED 30293SCLAR 30290YELLO 30293MOBBLUE 30290GREY 30288BLUE 30293MOBSCLAR 30280L 30280SCLAR 30289MOBGREY 30292RED 30283BLUE 30287YELLO 30289MOBRED 30281YELLO 30284RED 30284BLUE 30290MOBGREY 30289YELLO 30280RED 30283RED 30293RED 30289MOBYELLO 30290L 30293BLUE 30292MOBBLUE 30292YELLO 30290SCLAR 30290MOBYELLO 30282BLUE 30288MOBGREY 30290RED 30288SCLAR 30282L 30292MOBSCLAR 30286YELLO 30288MOBYELLO 30290MOBBLUE 30292GREY 30288MOBSCLAR 30290BLUE 30282YELLO 30283SCLAR 30288MOBRED 30287GREY
75855-168CS 424.58 USD
75855-168 75855-124 75855-202 75855-126 75855-204 75855-920 75855-128 75855-206 75855-922 75855-208 75855-924 75855-926 75856-048 75855-928 75856-046 75856-052 75856-050 75855-170 75855-094 75855-172 75855-096 75855-174 75855-130 75855-098 75855-132 75855-176 75855-210 75855-134 75855-178 75855-234 75855-114 75855-158 75855-116 75855-118 75855-918 75855-160 75855-162 75855-164 75855-120 75855-122 75855-166 75855-200 75855-102 75855-146 75855-224 75855-148 75855-104 75855-940 75855-226 75855-820 75855-106 75855-822 75855-228 75855-108 75855-824 75855-826 75855-190 75855-192 75855-150 75855-194 75855-152 75855-196 75855-230 75855-110 75855-154 75855-198 75855-232 75855-112 75855-156 75855-212 75855-136 75855-214 75855-930 75855-138 75855-216 75855-932 75855-218 75855-934 75855-936 75855-938 75855-180 75855-182 75855-184 75855-140 75855-186 75855-142 75855-220 75855-144 75855-188 75855-100 75855-222
Super-Size AkroBins, Akro-Mils
Boxes Dispenser Boxes
Super-Size AkroBins offer huge storage capacity and the versatility required to make the most of available space.
  • Reinforced side ribs add strength and prevent spreading when stacked
  • Large label area on front of bin
  • Hinged lid covers and protects while allowing access to contents, even when bins are stacked
  • Made in the USA
These heavy-duty plastic storage bins save space by securely stacking atop each other, and sitting on wire shelving and steel shelving, or steel carts and dollies. Super-Size AkroBins are resistant to weak acids and alkalis. Sturdy, one-piece construction is water, rust and corrosion-proof.
Accessories information: Keep items separated in bin using dividers, provides additional storage options.