Rabiner Neurohammer, OR Grade, Sklar
Supplier: Sklar
06-3290
10194-172EA 168.37 USD
10194-172
Rabiner Neurohammer, OR Grade, Sklar
Mallets and Hammers
Sklar's Rabiner Neurological Hammer is an altered version of the Babinsky Hammer, that features a hinge and circular rotating bumper so the head of the instrument can be used parallel or perpendicular to the handle
  • Made of stainless steel
Censitrac Ready products are industry standard.
This hammer also comes with a brush and needle, both of which screw into the end of the hollow handle.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.