1,2,4,5-Tetraaminobenzene tetrahydrochloride, Technical Grade

Supplier: Pfaltz & Bauer
Warning

Synonyms: Benzene-1,2,4,5-tetramine 4HClBenzene-1,2,4,5-tetrayltetraamine tetrahydrochloride1,2,4,5-Tetraaminobenzene tetrahydrochloride1,2,4,5-Benzenetetramine, tetrahydrochloride1,2,4,5-Benzenetetramine tetrahydrochlorideY15Benzene-1,2,4,5-tetraamine tetrahydrochloride

T01680-500MG
101184-311EA 820.32 USD
101184-311
1,2,4,5-Tetraaminobenzene tetrahydrochloride, Technical Grade
1,2,4,5-Tetraaminobenzene tetrahydrochloride
Formula: C₆H₂(NH₂)₄·4HCl
MW: 284.006 g/mol
Melting Pt: 300 °C
MDL Number: MFCD00012970
CAS Number: 4506-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR