n-Hexane ≥95%, CHROMASOLV™ for HPLC, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Hexyl hydride, Hexanen, Hexane

270504-6X1L 270504-4X4L
BJ270504-6X1LCS 835.11 USD
BJ270504-6X1L BJ270504-4X4L
n-Hexane ≥95%, CHROMASOLV™ for HPLC, Riedel-de Haën™
n-Hexane

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency


Beilstein Number: 1730733

Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 69
Melting Pt: -94.3
Density: 0.659…0.673 g/cm³ (20 °C)
Flash Pt: –26 °C
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3 II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity (IR) complying
Assay (GC) Min. 95.0 %
Refractive index (n 20/D) 1.373 - 1.377
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.020 %
Absorbance at 195 nm Max. 1.0
Absorbance at 210 nm Max. 0.30
Absorbance at 220 nm Max. 0.10
Absorbance at 230 nm Max. 0.05
Absorbance at 240 nm Max. 0.02
Absorbance at 250 nm Max. 0.01
Absorbance at 400 nm Max. 0.01
Vapor Pressure 256 mmHg ( 37.7 °C)
Vapor Density ~3 (vs air)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR