Acetonitrile ≥99.9% (by GC), CHROMASOLV™ for HPLC, for UV spectrophotometry, Riedel-de Haën™
Supplier: Honeywell Research Chemicals
|
Danger
|
Synonyms:
Methane, cyano-
, Ethyl nitrile
, Methyl cyanide
, Ethanoic acid nitrile
, Methanecarbonitrile
, Ethanenitrile
, Cyanomethane
, ACN
34888-6X1L
34888-1L
34888-2.5L
34888-4X4L
34888-4X2.5L
BJ34888-2.5LEA
428.36
USD
BJ34888-2.5L
BJ34888-4X2.5L
BJ34888-6X1L
BJ34888-4X4L
BJ34888-1L
Acetonitrile ≥99.9% (by GC), CHROMASOLV™ for HPLC, for UV spectrophotometry, Riedel-de Haën™
Acetonitrile
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Beilstein Number: 741857
Formula:
H₃CCN MW: 41.05 g/mol Boiling Pt: 82 °C (1.013 hPa) Melting Pt: -46 °C Density: 0.7857 g/cm³ (20 °C) Flash Pt: 6 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001878 CAS Number: 75-05-8 EINECS: 200-835-2 UN: 1648 ADR: 3,II |
Specification Test Results
Assay (GC) | Min. 99.9 % |
Non-volatile matter | Max. 0.0005 % |
Water (Karl Fischer) | Max. 0.02 % |
Free acid (as CH₃COOH) | Max. 0.002 % |
Free alkali (as NH₃) | Max. 0.0002 % |
Transmittance at 195 nm | Min. 65 % |
Transmittance at 200 nm | Min. 88 % |
Transmittance at 210 nm | Min. 93 % |
Transmittance at 235 nm | Min. 99 % |
Absorbance (JP)(Val.Ref) | complying |
Fluorescence (chinin) at 254 nm | Max. 1 ppb |
Fluorescence (chinin) at 365 nm | Max. 0.5 ppb |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...