Dichloromethane ≥99.8% (by GC) stabilised, CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Research Chemicals
|
Warning
|
Synonyms:
Methane dichloride, Methylene chloride, Methylene chloride
Stabilization:
Stabilised with amylene
34488-6X1L
34488-4X4L
34488-4X2.5L
BJ34488-1LCS
456.77
USD
BJ34488-1L
BJ34488-4L
BJ34488-2.5L
Dichloromethane ≥99.8% (by GC) stabilised, CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Dichloromethane
Formula:
CH₂Cl₂ MW: 84.93 g/mol Boiling Pt: 39.8 Melting Pt: -95 Density: 1.32…1.33 g/cm³ (20 °C) (lit.) Storage Temperature: Ambient |
MDL Number:
MFCD00000881 CAS Number: 75-09-2 EINECS: 200-838-9 UN: 1593 ADR: 6.1 III Merck Index: 12,06140 |
|
Specification Test Results
Assay (GC) | Min. 99.8 % |
Non-volatile matter | Max. 0.0005 % |
Water (Karl Fischer) | Max. 0.02 % |
Suitability for residue analysis | Max. 5 ng/l |
Contains | ~25 mg/L amylene as stabilizer |
Vapor Pressure | 6.83 psi ( 20 °C) |
Refractive Index | n20/D 1.424 (lit.) |
Explosion Limit | 22 % |
Vapor Density | 2.9 (vs air) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...