Celite® 545, filter aid ≥99% ≤125.30 μm (71% (acc. Cilas)), treated with sodium carbonate, flux calcined, Fluka™

Supplier: Honeywell Research Chemicals

Synonyms: DE, Diatomaceous earth, flux-calcined

22140-1KG 22140-5KG
BJ22140-1KGEA 130 USD
BJ22140-1KG BJ22140-5KG
Celite® 545, filter aid ≥99% ≤125.30 μm (71% (acc. Cilas)), treated with sodium carbonate, flux calcined, Fluka™
Celite® 545, filter aid
Formula: SiO₂
Storage Temperature: Ambient
MDL Number: MFCD00132803
CAS Number: 68855-54-9
EINECS: 272-489-0

Order Now

Specification Test Results

Composition Fe₂O₃, 0.46%
pH (10 % susp., 25°C) Min. 8.5
Loss on ignition (900°C) Max. 0.3 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR