Sodium hydroxide 1.0 M NaOH (1.0N) Volumetric, Fluka™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

319511-1L 319511-2L 319511-500ML 319511-6X1L
BJ319511-1LEA 36 USD
BJ319511-1L BJ319511-2L BJ319511-500ML BJ319511-6X1L
Sodium hydroxide 1.0 M NaOH (1.0N) Volumetric, Fluka™
Sodium hydroxide
Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390
Melting Pt: 323
Density: 1.04 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1824
ADR: 8 II
Merck Index: 14,08627

Order Now

Specification Test Results

Assay 0.950 - 1.050 N
Carbonate (as Na₂CO₃) Max. 0.03 N
Traceable to NIST SRM effective
Vapor Pressure 3 mmHg ( 37 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR