Fernbach Culture Flasks, without Baffles, Chemglass
Supplier: Chemglass
CLS-2020-30 CLS-2020-11 CLS-2020-10
30623-124EA 308.4 USD
30623-124 30623-128 30623-122
Fernbach Culture Flasks, without Baffles, Chemglass
Flasks Culture Flasks
These flasks are designed for culturing organisms that require a large surface area to volume ratio.
Available with a plain 59 mm ID neck, 38 mm OD Delong style neck or GL-45 screw neck supplied with a vented cap.