FlyFectIN™ Transfection Reagent, OZ Biosciences
Supplier: OZ Biosciences
FF50500 FF51000 FF55000
103258-448EA 217.51 USD
103258-448 103258-450 103258-452
FlyFectIN™ Transfection Reagent, OZ Biosciences
Transfection Reagents
FlyFectin Transfection Reagent is a powerful reagent based on the TEE-technology and specifically designed to obtain highly efficient and reproducible transfection of insect cells.
  • Dedicated to insect cells
  • Ideal for production of recombinant protein using Baculovirus expression system.
  • Non toxic
  • Serum compatible
  • Simple, Ready-to-use and Rapid
It is adapted to the delivery of all types of nucleic acids.
Delivery information: Shipped at room temperature
Packaging: Store at +4 °C