EcoTransfect™ Transfection Reagent, OZ Biosciences
Supplier: OZ Biosciences
ET10500 ET11000 ET13000
103258-396EA 138.92 USD
103258-396 103258-398 103258-400
EcoTransfect™ Transfection Reagent, OZ Biosciences
Transfection Reagents
EcoTransfect Transfection Reagent is an economical Lipofection reagent dedicated to the transfection of popular cell lines.
  • Economical
  • Good efficiency
  • Serum compatible
  • Simple, ready-to-use and rapid
It is the perfect solution to quickly analyze the biological activity of your nucleic acids, to perform routine transfection assays at low cost and to accomplish high throughput screening.
Delivery information: Shipped at room temperature
Packaging: Store at +4 °C