DreamFect™ Stem Transfection Reagent, OZ Biosciences
Supplier: OZ Biosciences
ST30500
ST31000
103258-392EA
267.89
USD
103258-392
103258-394
DreamFect™ Stem Transfection Reagent, OZ Biosciences
Transfection Reagents
DreamFect Stem Transfection Reagent is a powerful reagent allowing multipotent stem cells transfection with high efficiency and very low toxicity.
- High transfection efficiency for multipotent Stem cells
- Minimized toxicity due to reagent biodegradability and low DNA amount required
- Cell phenotype and differentiation potential are not affected
- Serum Compatible
- Simple, Ready-to-use and rapid (no specific buffer)
Delivery information: Shipped at room temperature
Packaging: Store at -20 °C