Pro-DeliverIN™ Transfection Reagent, OZ Biosciences
Supplier: OZ Biosciences
PI10100 PI10250 PI10500 PI11000
103258-302EA 224.88 USD
103258-302 103258-304 103258-306 103258-308
Pro-DeliverIN™ Transfection Reagent, OZ Biosciences
Transfection Reagents
Pro-DeliverIN Transfection Reagent is an innovative reagent allowing intracellular delivery of biologically active proteins.
  • Intracellular delivery of functionally active proteins
  • Highly efficient in many primary cells and cell lines
  • Serum compatible
  • Biodegradable and non toxic
  • Straightforward protocol and ready-to-use reagent
This lipid-based formulation is the first serum compatible reagent to deliver functional proteins into living cells. The proteins delivered inside cells retain their biological functions.
Delivery information: Shipped at room temperature
Packaging: Store at +4 °C