LipoMag™ Transfection Kit, OZ Biosciences
Supplier: OZ Biosciences
LM80500 LM80000 LM80250
103258-218EA 752.37 USD
103258-218 103258-454 103258-456
LipoMag™ Transfection Kit, OZ Biosciences
Transfection Reagents
LipoMag Transfection Kit associates our most efficient lipid-based transfection reagent, DreamFect Gold with CombiMag magnetic nanoparticles, the most versatile Magnetofection reagent.
  • Highest efficiency without toxicity
  • Superior transgene expression level than any other reagents
  • Enhancement of Dreamfect™ Gold efficiency and outperformed competitors
  • Biodegradable
  • Multipurpose (various types of nucleic acids)
  • Universal (cell lines and primary cells)
  • Simple, ready-to-use and rapid
  • Serum compatible
By taking advantage of the Lipofection and Magnetofection transfection techniques, LipoMag Kit reaches even higher transfection efficiency especially in primary cells while reducing toxicity.
Accessories information: This kit needs to be used with a magnetic plate
Delivery information: Shipped at room temperature
Packaging: Store DreamFect Gold -20°C and CombiMag at +4°C