DreamFect™ Gold Transfection Reagent, OZ Biosciences
Supplier: OZ Biosciences
DG80500
DG81000
DG85000
103258-212EA
267.89
USD
103258-212
103258-214
103258-216
DreamFect™ Gold Transfection Reagent, OZ Biosciences
Transfection Reagents
DreamFect Gold Transfection Reagent allows transfecting all types of nucleic acids with a very high efficiency.
- High efficiency
- Higher protein expression
DreamFect Gold delivers a large quantity of nucleic acids leading to higher protein expression compared to other transfection reagents.
Delivery information: Shipped at room temperature.
Packaging: Store at -20 °C