HEPES-2-[4-(2-Hydroxyethyl)-1-piperazinyl]-ethanesulfonic acid ≥99.5%, white crystalline powder for molecular biology

Supplier: Promega Corporation

H5302 H5303
PAH5302EA 108.37 USD
PAH5302 PAH5303
HEPES-2-[4-(2-Hydroxyethyl)-1-piperazinyl]-ethanesulfonic acid ≥99.5%, white crystalline powder for molecular biology
HEPES (4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid)

HEPES is a biological buffer that functions over a pH range of 6.8 to 8.2.

Formula: C₈H₁₈N₂O₄S
Storage Temperature: Ambient
MDL Number: MFCD00006158
CAS Number: 7365-45-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4cdc09c866676149941b93b2cac8d453175059a2&vid=0b09e5b84ea34a806443c3071fad63e15e4e3433&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White, crystalline powder
Purity ≥99.5%
Lead ≤5 ppm
Iron ≤5 ppm
Moisture ≤0.5%
pH at 25°C (1M) 5.0 – 6.5
A260 at 0.1M ≤0.05
A280 at 0.1M ≤0.04


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR