Chemical OSHA Danger Signs, Chlorine, National Marker
Supplier: ACCUFORM MANUFACTURING, INC
D15AB D15P D484AB D484PB D484RB
94008-508EA 59.76 USD
94008-508 94008-510 94008-512 94010-332 94010-334 94010-336
Chemical OSHA Danger Signs, Chlorine, National Marker
Signs
OSHA signs are the perfect way to alert your employees and guests when a hazard is present in your facility.
  • Ideal for indoor and outdoor labeling
  • Available in Alum, PS vinyl, Fiber glass and Rigid plastic (thick polystyrene) materials
  • .040 alum is weatherproof, non-corrosive and has baked-on enamel on both sides
  • .004 PS vinyl material has pressure sensitive adhesive backing
  • .095 fiber glass material is rigid, shatter-proof and dent resistant
  • .050 rigid plastic material has radius corners and pre-drilled corner mounting holes
Keep your facility safe, buy an OSHA regulated sign today and prevent an accident tomorrow. Each sign header reads "Danger".
Certifications: Comply with OSHA 29 CFR 1910.