Alcian Blue pH 2.5, Goblet Cell Histo Positive Control Slides, Newcomer Supply
Supplier: Newcomer Supply
4051A 4051B
75876-956BX 75.55 USD
75876-956 75876-958
Alcian Blue pH 2.5, Goblet Cell Histo Positive Control Slides, Newcomer Supply
Slides Microscopy Control Slides
Alcian blue pH 2.5, goblet cell control slides contain a section of positive staining small intestine.
  • Tissue type: (+) Small Intestine
  • All control tissue is sectioned on Superfrost® Plus slides
  • Staining validation technical memo included in each set
  • All tissues are formalin fixed, paraffin embedded FFPE
  • All slides are air dried
  • Control slides are cut at 4 µm, unless otherwise specified
  • Standard tissue placement is mid-high on slide
Newcomer supply alcian blue pH 2.5, goblet cell control slides are for the positive histochemical staining of gobletMore Product Information