Thyroid Transcription Factor (TTF-1) IHC Positive Control Slides, Newcomer Supply
Supplier: Newcomer Supply
3780A 3780B
75877-134BX 91.27 USD
75877-134 75877-136
Thyroid Transcription Factor (TTF-1) IHC Positive Control Slides, Newcomer Supply
Slides Microscopy Control Slides
Thyroid Transcription Factor (TTF-1) control slides contain sections of positive staining thyroid and negative staining myometrium.
  • Tissue type: (+) Thyroid / (-) Myometrium
  • All control tissue is sectioned on Superfrost® Plus slides
  • Staining validation technical memo included in each set
  • All tissues are formalin fixed, paraffin embedded FFPE
  • All slides are air dried
  • IHC slides are cut at 4 µm
  • Standard tissue placement is mid-high on slide
Newcomer supply Thyroid Transcription Factor (TTF-1) control slides are for the positive immunohistochemicalMore Product Information