S-100 IHC Positive Control Slides, Newcomer Supply

Supplier: Newcomer Supply
3740A 3740B
75876-922BX 91.27 USD
75876-922 75876-924
S-100 IHC Positive Control Slides, Newcomer Supply
S-100 control slides contain sections of positive staining melanoma and negative staining lung.

  • Tissue type: (+) Melanoma / (-) Lung
  • All control tissue is sectioned on Superfrost® Plus slides
  • Staining validation technical memo included in each set
  • All tissues are formalin fixed, paraffin embedded FFPE
  • All slides are air dried
  • IHC slides are cut at 4 µm
  • Standard tissue placement is mid-high on slide

Newcomer supply S-100 control slides are for the positive immunohistochemical staining of S-100, a ubiquitous protein expressed in a number of cells

Staining Results:
S-100 positive expression will stain brown cytoplasmic & nuclear staining.
Lung will stain negative.

These positive control slides are intended to be used to verify histological techniques and reagent reactivity. These slides are to be used for the qualitative purpose of determining positive or negative results, and are not intended to be used for any quantitative purpose.

Its demonstration is of great value in the identification of several neoplasms, particularly melanomas.

Ordering information: All controls are cut in serial sections.
10 set = 10 unstained slides and 1 stained slide.
98 set = 98 unstained slides and 2 stained slides. (First & Last Slides are Stained)
Reactivity of IHC control slides are guaranteed for one year from the date of receipt and should be revalidated after one year to verify continued reactivity.
Please note the following: Allow 2-3 weeks for delivery for large sets. In human tissue unless otherwise indicated.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR